2-Chloro-3-(2-chloroethyl)-6-ethoxyquinoline

Lieferant: Apollo Scientific

OR307895-1G
APOSOR307895-1GEA 335 EUR
APOSOR307895-1G
2-Chloro-3-(2-chloroethyl)-6-ethoxyquinoline
2-Chloro-3-(2-chloroethyl)-6-ethoxyquinoline
Formel: C₁₃H₁₃Cl₂NO
Molecular Weight: 270,154 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787619
CAS-Nummer: 948294-49-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR