2-Chloro-3-(2-chloroethyl)-7-bromoquinoline

Lieferant: Apollo Scientific

OR307896-250MG
APOSOR307896-250MGEA 177 EUR
APOSOR307896-250MG
2-Chloro-3-(2-chloroethyl)-7-bromoquinoline
2-Chloro-3-(2-chloroethyl)-7-bromoquinoline
Formel: C₁₁H₈BrCl₂N
Molecular Weight: 304,998 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787622
CAS-Nummer: 948294-53-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR