3-(2-Chloroethyl)-2,7-dichloroquinoline

Lieferant: Apollo Scientific

OR307897-1G
APOSOR307897-1GEA 359 EUR
APOSOR307897-1G
3-(2-Chloroethyl)-2,7-dichloroquinoline
3-(2-Chloroethyl)-2,7-dichloroquinoline
Formel: C₁₁H₈Cl₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD09787623
CAS-Nummer: 948294-54-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR