2-Chloro-3-(3-chloropropyl)-6-methoxyquinoline

Lieferant: Apollo Scientific

OR307900-1G
APOSOR307900-1GEA 359 EUR
APOSOR307900-1G
2-Chloro-3-(3-chloropropyl)-6-methoxyquinoline
2-Chloro-3-(3-chloropropyl)-6-methoxyquinoline
Formel: C₁₃H₁₃Cl₂NO
Molecular Weight: 270.154 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787635
CAS-Nummer: 159383-57-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR