2-Chlor-3-(chlormethyl)-5,7-dimethylchinolin

Lieferant: Apollo Scientific

OR307904-1G
APOSOR307904-1GEA 343 EUR
APOSOR307904-1G
2-Chlor-3-(chlormethyl)-5,7-dimethylchinolin
2-Chloro-3-(chloromethyl)-5,7-dimethylquinoline
Formel: C₁₂H₁₁Cl₂N
Lagertemperatur: Raumtemperatur
CAS-Nummer: 948290-59-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR