2-Chlor-3-(chlormethyl)-6,7-dimethylchinolin

Lieferant: Apollo Scientific

OR307905-1G
APOSOR307905-1GEA 488 EUR
APOSOR307905-1G
2-Chlor-3-(chlormethyl)-6,7-dimethylchinolin
2-Chloro-3-(chloromethyl)-6,7-dimethylquinoline
Formel: C₁₂H₁₁Cl₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD09787653
CAS-Nummer: 182052-67-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR