2-Chlor-3-(chlormethyl)-6-ethoxychinolin

Lieferant: Apollo Scientific

OR307907-1G
APOSOR307907-1GEA 343 EUR
APOSOR307907-1G
2-Chlor-3-(chlormethyl)-6-ethoxychinolin
2-Chloro-3-(chloromethyl)-6-ethoxyquinoline
Formel: C₁₂H₁₁Cl₂NO
Lagertemperatur: Raumtemperatur
CAS-Nummer: 948290-90-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR