3-(Chlormethyl)-2,7-dichlorchinolin
Lieferant: Apollo Scientific
OR307908-1G
APOSOR307908-1GEA
343
EUR
APOSOR307908-1G
3-(Chlormethyl)-2,7-dichlorchinolin
3-(Chloromethyl)-2,7-dichloroquinoline
Formel:
C₁₀H₆Cl₃N Lagertemperatur: Raumtemperatur |
CAS-Nummer:
137776-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...