2-Chlor-3-(chlormethyl)-7-methoxychinolin
Lieferant: Apollo Scientific
OR307909-1G
APOSOR307909-1GEA
335
EUR
APOSOR307909-1G
2-Chlor-3-(chlormethyl)-7-methoxychinolin
2-Chloro-3-(chloromethyl)-7-methoxyquinoline
Formel:
C₁₁H₉Cl₂NO Lagertemperatur: Raumtemperatur |
CAS-Nummer:
73863-49-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...