2-Chloro-5,7-dimethylquinoline-3-carbonitrile

Lieferant: Apollo Scientific

OR307911-1G OR307911-5G
APOSOR307911-1GEA 0 EUR
APOSOR307911-1G APOSOR307911-5G
2-Chloro-5,7-dimethylquinoline-3-carbonitrile
2-Chloro-5,7-dimethylquinoline-3-carbonitrile
Formel: C₁₂H₉ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08899072
CAS-Nummer: 917747-10-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR