2,6-Dichloroquinoline-3-carbonitrile

Lieferant: Apollo Scientific

OR307917-1G
APOSOR307917-1GEA 359 EUR
APOSOR307917-1G
2,6-Dichloroquinoline-3-carbonitrile
2,6-Dichloroquinoline-3-carbonitrile
Formel: C₁₀H₄Cl₂N₂
Molecular Weight: 223,058 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787669
CAS-Nummer: 948291-61-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR