2-Chloro-6-ethylquinoline-3-carbonitrile

Lieferant: Apollo Scientific

OR307921-1G
APOSOR307921-1GEA 557 EUR
APOSOR307921-1G
2-Chloro-6-ethylquinoline-3-carbonitrile
2-Chloro-6-ethylquinoline-3-carbonitrile
Formel: C₁₂H₉ClN₂
Molecular Weight: 216.666 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03047545
CAS-Nummer: 498548-90-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR