2-Chloro-8-ethyl-quinoline-3-carbaldehyde

Lieferant: Apollo Scientific

OR307927-1G OR307927-5G
APOSOR307927-1GEA 301 EUR
APOSOR307927-1G APOSOR307927-5G
2-Chloro-8-ethyl-quinoline-3-carbaldehyde
2-Chloro-8-ethyl-quinoline-3-carbaldehyde
Formel: C₁₂H₁₀ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD02587932
CAS-Nummer: 335196-05-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR