4,7-Dichlor-2,8-dimethylchinolin

Lieferant: Apollo Scientific

OR307933-5G
APOSOR307933-5GEA 163 EUR
APOSOR307933-5G
4,7-Dichlor-2,8-dimethylchinolin
4,7-Dichloro-2,8-dimethylquinoline
Formel: C₁₁H₉Cl₂N
Molecular Weight: 226.1000 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00272342
CAS-Nummer: 21728-15-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR