4,7-Dichlor-2-methylchinolin ≥97%

Lieferant: Apollo Scientific
Gefahr

OR307934-250MG
APOSOR307934-250MGEA 84.1 EUR
APOSOR307934-250MG
4,7-Dichlor-2-methylchinolin ≥97%
4,7-Dichloro-2-methylquinoline
Formel: C₁₀H₇Cl₂N
Lagertemperatur: Kühlschrank
CAS-Nummer: 50593-69-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR