4,8-Dichlor-2,6-dimethylchinolin
Lieferant: Apollo Scientific
OR307935-1G
APOSOR307935-1GEA
383
EUR
APOSOR307935-1G
4,8-Dichlor-2,6-dimethylchinolin
4,8-Dichloro-2,6-dimethylquinoline
Formel:
C₁₁H₉Cl₂N Lagertemperatur: Raumtemperatur |
CAS-Nummer:
203626-46-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...