4-Amino-5,7-dimethylchinolin

Lieferant: Apollo Scientific

OR307938-1G
APOSOR307938-1GEA 393 EUR
APOSOR307938-1G
4-Amino-5,7-dimethylchinolin
4-Amino-5,7-dimethylquinoline
Formel: C₁₁H₁₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09787704
CAS-Nummer: 948292-64-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR