4-Amino-6,7-dichlorchinolin
Lieferant: Apollo Scientific
OR307939-1G
APOSOR307939-1GEA
426
EUR
APOSOR307939-1G
4-Amino-6,7-dichlorchinolin
4-Amino-6,7-dichloroquinoline
Formel:
C₉H₆Cl₂N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08063229 CAS-Nummer: 948292-94-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...