4-Amino-6,7-dimethylchinolin

Lieferant: Apollo Scientific

OR307940-1G
APOSOR307940-1GEA 417 EUR
APOSOR307940-1G
4-Amino-6,7-dimethylchinolin
4-Amino-6,7-dimethylquinoline
Formel: C₁₁H₁₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09787710
CAS-Nummer: 948292-99-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR