4-Amino-6,8-dichlorchinolin

Lieferant: Apollo Scientific

Synonyme: 6,8-Dichloroquinolin-4-amine

OR307941-1G
APOSOR307941-1GEA 568 EUR
APOSOR307941-1G
4-Amino-6,8-dichlorchinolin
4-Amino-6,8-dichloroquinoline
Formel: C₉H₆Cl₂N₂
Molecular Weight: 213,633 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08063227
CAS-Nummer: 929339-40-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR