4-Amino-6-chloro-8-methylquinoline
Lieferant: Apollo Scientific
OR307943-1G
APOSOR307943-1GEA
0
EUR
APOSOR307943-1G
4-Amino-6-chloro-8-methylquinoline
4-Amino-6-chloro-8-methylquinoline
Formel:
C₁₀H₉ClN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09787713 CAS-Nummer: 948293-08-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...