4-Amino-6-ethoxyquinoline
Lieferant: Apollo Scientific
OR307944-1G
APOSOR307944-1GEA
383
EUR
APOSOR307944-1G
4-Amino-6-ethoxyquinoline
4-Amino-6-ethoxyquinoline
Formel:
C₁₁H₁₂N₂O Molecular Weight: 188,226 g/mol Schmelzpunkt: 195 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09787715 CAS-Nummer: 948293-13-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...