4-Amino-6-ethylquinoline

Lieferant: Apollo Scientific

OR307945-1G
APOSOR307945-1GEA 375 EUR
APOSOR307945-1G
4-Amino-6-ethylquinoline
4-Amino-6-ethylquinoline
Formel: C₁₁H₁₂N₂
Molecular Weight: 172,226 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787717
CAS-Nummer: 948293-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR