4-Amino-6-methylchinolin

Lieferant: Apollo Scientific

OR307946-1G
APOSOR307946-1GEA 0 EUR
APOSOR307946-1G
4-Amino-6-methylchinolin
4-Amino-6-methylquinoline
Formel: C₁₀H₁₀N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06761164
CAS-Nummer: 874589-77-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR