4-Amino-7,8-dichlorchinolin

Lieferant: Apollo Scientific

OR307947-1G
APOSOR307947-1GEA 359 EUR
APOSOR307947-1G
4-Amino-7,8-dichlorchinolin
4-Amino-7,8-dichloroquinoline
Formel: C₉H₆Cl₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09787718
CAS-Nummer: 948293-25-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR