4-Amino-7-bromchinolin

Lieferant: Apollo Scientific

Synonyme: 7-Bromoquinolin-4-amine

OR307949-1G
APOSOR307949-1GEA 451 EUR
APOSOR307949-1G
4-Amino-7-bromchinolin
4-Amino-7-bromoquinoline
Formel: C₉H₇BrN₂
Molecular Weight: 223,0693 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06411037
CAS-Nummer: 65340-74-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR