4-Amino-7-chloro-8-methylquinoline

Lieferant: Apollo Scientific

OR307951-1G
APOSOR307951-1GEA 359 EUR
APOSOR307951-1G
4-Amino-7-chloro-8-methylquinoline
4-Amino-7-chloro-8-methylquinoline
Formel: C₁₀H₉ClN₂
Molecular Weight: 192,645 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787721
CAS-Nummer: 948293-41-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR