4-Amino-7-methoxy-2-methylchinolin
Lieferant: Apollo Scientific
OR307953-1G
APOSOR307953-1GEA
451
EUR
APOSOR307953-1G
4-Amino-7-methoxy-2-methylchinolin
4-Amino-7-methoxy-2-methylquinoline
Formel:
C₁₁H₁₂N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD09787723 CAS-Nummer: 66735-41-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...