4-Amino-8-chloro-6-methylquinoline

Lieferant: Apollo Scientific

OR307956-1G
APOSOR307956-1GEA 0 EUR
APOSOR307956-1G
4-Amino-8-chloro-6-methylquinoline
4-Amino-8-chloro-6-methylquinoline
Formel: C₁₀H₉ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09787725
CAS-Nummer: 948293-57-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR