4-Chlor-6,7-dimethylchinolin

Lieferant: Apollo Scientific
Achtung

OR307964-1G
APOSOR307964-1GEA 0 EUR
APOSOR307964-1G
4-Chlor-6,7-dimethylchinolin
4-Chloro-6,7-dimethylquinoline
Formel: C₁₁H₁₀ClN
Schmelzpunkt: 95…96 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 861038-79-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR