4-Chlor-6,8-dimethylchinolin
Lieferant: Apollo Scientific
OR307965-1G
APOSOR307965-1GEA
95.7
EUR
APOSOR307965-1G
4-Chlor-6,8-dimethylchinolin
4-Chloro-6,8-dimethylquinoline
Formel:
C₁₁H₁₀ClN Lagertemperatur: Raumtemperatur |
MDL:
MFCD00087699 CAS-Nummer: 196803-72-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...