4-Chlor-7-methylchinolin

Lieferant: Apollo Scientific
Achtung

OR307970-1G
APOSOR307970-1GEA 551 EUR
APOSOR307970-1G
4-Chlor-7-methylchinolin
4-Chloro-7-methylquinoline
Formel: C₁₀H₈ClN
Molecular Weight: 177,630 g/mol
Schmelzpunkt: 27…28 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09752823
CAS-Nummer: 63136-61-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR