4-Hydroxyquinoline-3-carboxylic acid
Lieferant: Apollo Scientific
OR307982-5G
APOSOR307982-5GEA
75.4
EUR
APOSOR307982-5G
4-Hydroxyquinoline-3-carboxylic acid
4-Hydroxyquinoline-3-carboxylic acid
Formel:
C₁₀H₇NO₃ Molecular Weight: 189.167 g/mol Schmelzpunkt: 268…273 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00598887 CAS-Nummer: 34785-11-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...