Ethyl-4-hydroxychinolin-3-carboxylat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Hydroxyquinoline-3-carboxylic acid ethyl ester, Ethyl 4-hydroxyquinoline-3-carboxylate
OR307983-250MG
APOSOR307983-250MGEA
21.9
EUR
APOSOR307983-250MG
Ethyl-4-hydroxychinolin-3-carboxylat 98%
Ethyl-4-hydroxychinolin-3-carboxylat
Formel:
C₁₂H₁₁NO₃ Molecular Weight: 217,2206 g/mol Schmelzpunkt: 271 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173406 CAS-Nummer: 26892-90-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...