5,7-Dichlor-2-methylchinolin

Lieferant: Apollo Scientific

OR307984-1G
APOSOR307984-1GEA 393 EUR
APOSOR307984-1G
5,7-Dichlor-2-methylchinolin
5,7-Dichloro-2-methylquinoline
Formel: C₁₀H₇Cl₂N
Lagertemperatur: Raumtemperatur
CAS-Nummer: 26933-09-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR