5,7-Dichloro-4-hydrazinoquinoline hydrochloride

Lieferant: Apollo Scientific

OR307985-1G
APOSOR307985-1GEA 451 EUR
APOSOR307985-1G
5,7-Dichloro-4-hydrazinoquinoline hydrochloride
5,7-Dichloro-4-hydrazinoquinoline hydrochloride
Formel: C₉H₇Cl₂N₃·ClH
Molecular Weight: 264,539 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787762
CAS-Nummer: 1172490-52-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR