5,7-Dichloroquinoline-3-carboxylic acid
Lieferant: Apollo Scientific
OR307986-1G
APOSOR307986-1GEA
368
EUR
APOSOR307986-1G
5,7-Dichloroquinoline-3-carboxylic acid
5,7-Dichloroquinoline-3-carboxylic acid
Formel:
C₁₀H₅Cl₂NO₂ Molecular Weight: 242,058 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09787766 CAS-Nummer: 948293-83-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...