5,8-Dichloro-4-hydroxyquinoline-3-carboxylic acid

Lieferant: Apollo Scientific

OR307989-1G
APOSOR307989-1GEA 351 EUR
APOSOR307989-1G
5,8-Dichloro-4-hydroxyquinoline-3-carboxylic acid
5,8-Dichloro-4-hydroxyquinoline-3-carboxylic acid
Formel: C₁₀H₅Cl₂NO₃
Molecular Weight: 258,058 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787773
CAS-Nummer: 35973-26-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR