5,8-Dimethylquinoline-3-carboxylic acid

Lieferant: Apollo Scientific

OR307990-1G
APOSOR307990-1GEA 1270 EUR
APOSOR307990-1G
5,8-Dimethylquinoline-3-carboxylic acid
5,8-Dimethylquinoline-3-carboxylic acid
Formel: C₁₂H₁₁NO₂
Molecular Weight: 201,221 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787777
CAS-Nummer: 763893-29-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR