5-Chlor-2,8-dimethylchinolin
Lieferant: Apollo Scientific
OR307991-1G
APOSOR307991-1GEA
417
EUR
APOSOR307991-1G
5-Chlor-2,8-dimethylchinolin
5-Chloro-2,8-dimethylquinoline
Formel:
C₁₁H₁₀ClN Molecular Weight: 191,6600 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09787781 CAS-Nummer: 186670-43-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...