5-Chloro-8-methylquinoline-3-carboxylic acid

Lieferant: Apollo Scientific

OR307994-1G
APOSOR307994-1GEA 335 EUR
APOSOR307994-1G
5-Chloro-8-methylquinoline-3-carboxylic acid
5-Chloro-8-methylquinoline-3-carboxylic acid
Formel: C₁₁H₈ClNO₂
Molecular Weight: 221,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787789
CAS-Nummer: 948294-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR