6,7-Dimethyl-4-hydroxyquinoline

Lieferant: Apollo Scientific

OR307996-1G
APOSOR307996-1GEA 325 EUR
APOSOR307996-1G
6,7-Dimethyl-4-hydroxyquinoline
6,7-Dimethyl-4-hydroxyquinoline
Formel: C₁₁H₁₁NO
Molecular Weight: 173,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00272457
CAS-Nummer: 185437-33-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR