6,7-Dimethylquinoline-3-carboxylic acid

Lieferant: Apollo Scientific

OR307998-1G
APOSOR307998-1GEA 368 EUR
APOSOR307998-1G
6,7-Dimethylquinoline-3-carboxylic acid
6,7-Dimethylquinoline-3-carboxylic acid
Formel: C₁₂H₁₁NO₂
Molecular Weight: 201,221 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787801
CAS-Nummer: 948294-50-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR