tert-Butyl [4-(methylamino)butyl]carbamate

Lieferant: Apollo Scientific

OR308003-1G OR308003-5G
APOSOR308003-1GEA 0 EUR
APOSOR308003-1G APOSOR308003-5G
tert-Butyl [4-(methylamino)butyl]carbamate
tert-Butyl [4-(methylamino)butyl]carbamate
Formel: C₁₀H₂₂N₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD06808587
CAS-Nummer: 874831-66-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR