2-Hydroxymethyloxetane
Lieferant: Apollo Scientific
Synonyme:
2-Oxetanemethanol, Oxetan-2-ylmethanol, 2,4-Epoxybutanol
OR308011-1G
APOSOR308011-1GEA
98.6
EUR
APOSOR308011-1G
2-Hydroxymethyloxetane
2-Hydroxymethyloxetane
Formel:
C₄H₈O₂ Molecular Weight: 88,11 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00142933 CAS-Nummer: 61266-70-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...