3-[(Phenylmethyl)amino]-3-oxetanemethanamine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Aminomethyl)-N-benzyloxetan-3-amine

OR308012-1G
APOSOR308012-1GEA 509 EUR
APOSOR308012-1G
3-[(Phenylmethyl)amino]-3-oxetanemethanamine 97%
3-[(Phenylmethyl)amino]-3-oxetanemethanamine
Formel: C₁₁H₁₆N₂O
Molecular Weight: 192,26 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD17168407
CAS-Nummer: 1021392-83-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR