tert-Butyl 3-ethynylazetidine-1-carboxylate

Lieferant: Apollo Scientific

OR308016-1G
APOSOR308016-1GEA 87 EUR
APOSOR308016-1G
tert-Butyl 3-ethynylazetidine-1-carboxylate
tert-Butyl 3-ethynylazetidine-1-carboxylate
Formel: C₁₀H₁₅NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD17016080
CAS-Nummer: 287193-01-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR