2-(1-tert-Butoxycarbonylazetidin-3-yl)acetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-[1-Boc-azetidin-3-yl]acetic acid, 2-[1-(tert-Butoxycarbonyl)azetidin-3-yl]acetic acid

OR308024-25G
APOSOR308024-25GEA 89.9 EUR
APOSOR308024-25G
2-(1-tert-Butoxycarbonylazetidin-3-yl)acetic acid
2-(1-tert-Butoxycarbonylazetidin-3-yl)acetic acid
Formel: C₁₀H₁₇NO₄
Molecular Weight: 215.25 g/mol
Siedepunkt: 341.6 °C/ 760mmHg
Lagertemperatur: Kühlschrank
MDL: MFCD02179028
CAS-Nummer: 183062-96-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR