1-[(tert-Butoxy)carbonyl]-3-methylazetidine-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR308025-1G
APOSOR308025-1GEA 85.7 EUR
APOSOR308025-1G
1-[(tert-Butoxy)carbonyl]-3-methylazetidine-3-carboxylic acid
1-[(tert-Butoxy)carbonyl]-3-methylazetidine-3-carboxylic acid
Formel: C₁₀H₁₇NO₄
Lagertemperatur: Kühlschrank
MDL: MFCD12406581
CAS-Nummer: 887591-62-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR