1-Methylazetidine-3-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

OR308026-250MG
APOSOR308026-250MGEA 21.9 EUR
APOSOR308026-250MG
1-Methylazetidine-3-carboxylic acid 97%
1-Methylazetidine-3-carboxylic acid
Formel: C₅H₉NO₂
Molecular Weight: 115,1300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11520621
CAS-Nummer: 875629-26-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR